missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRBC1 Antibody (JOVI.1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50355APC
This item is not returnable.
View return policy
Description
TRBC1 Monoclonal antibody specifically detects TRBC1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF
Specifications
TRBC1 | |
Monoclonal | |
APC | |
BV05S1J2.2, TCRBC1, TRBC1 T cell receptor beta constant 1 | |
Human HA1.7 TCR beta chain expressed on transgenic mouse cells. | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
28639 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
JOVI.1 | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TRBC1 Antibody (JOVI.1), Allophycocyanin, Novus Biologicals™ > 0.1 mL, APC
Spot an opportunity for improvement?Share a Content Correction