Learn More
uvex Trend Black/Blue PU 69358 Boot - Width 11
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
107.00 EUR
Specifications
Color | Black, Blue |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17283721
|
uvex
6935838 |
38 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17293721
|
uvex
6935839 |
39 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17203731
|
uvex
6935840 |
40 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17213731
|
uvex
6935841 |
41 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17223731
|
uvex
6935842 |
42 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17233731
|
uvex
6935843 |
43 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17243731
|
uvex
6935844 |
44 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17253731
|
uvex
6935845 |
45 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17263731
|
uvex
6935846 |
46 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17273731
|
uvex
6935847 |
47 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17283731
|
uvex
6935848 |
48 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17293731
|
uvex
6935849 |
49 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17203741
|
uvex
6935850 |
50 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17213741
|
uvex
6935851 |
51 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
17223741
|
uvex
6935852 |
52 |
107.00 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
Description
Uvex 2 Trend Boot 69358 S2 SRC
- Sporty S2 safety boot with wide fit
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Smooth-running, dirt and heat-resistant lacing system
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Uvex Climazone technology lining, upper and removable comfortable climatic insole
- Moisture transport system helps maintain foot temperature
- Soft padding collar and dust tongue
- Removable and anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
- Article No. 69358
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 38 to 52
Specifications
Black, Blue | |
Dual-density polyurethane (PU/PU) | |
Reusable | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Steel | |
EN ISO 20345:2011 | |
Standard Width | |
Yes | |
S2 | |
ESD Safety Shoe |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
uvex Trend Black/Blue PU 69358 Boot - Width 11