Learn More
Trovirdine, MedChemExpress
MedChemExpress Trovirdine inhibits HIV-1 RT with an IC50 of 7 nM when employing heteropolymeric primer/template (oligo-DNA/ribosomal RNA)and dGTP as substrate.
Brand: Medchem Express HY-15349
This item is not returnable.
View return policy
Description
Trovirdine inhibits HIV-1 RT with an IC50 of 7 nM when employing heteropolymeric primer/template (oligo-DNA/ribosomal RNA)and dGTP as substrate.Specifications
Trovirdine | |
White | |
C13H13BrN4S | |
LY300046 | |
S=C(NCCC1=NC=CC=C1)NC2=NC=C(Br)C=C2 | |
337.24 | |
Research | |
COVID-19-anti-virus |
149488-17-5 | |
Research | |
10 mg | |
DMSO : 100 mg/mL (296.52 mM; Need ultrasonic) | |
337.24 | |
98.59% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Trovirdine, MedChemExpress