missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Works Calibration Certificate - Factory 160°C

Factory calibration certificate +160°C

Brand:  Memmert™ WTB35D00123

 View more versions of this product

Product Code. 17150385

  • 344.00 EUR / Each

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Accessory
  • Factory calibration certificate
  • 160°C
Specifications

Specifications

WTB35
Works Calibration Certificate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Memmert™ Works Calibration Certificate - Factory 160°C > WTB35

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.