Learn More
Memmert™ Works Calibration Certificate - Factory 160°C
Factory calibration certificate +160°C
344.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17120305
|
Memmert™
WTB11D00123 |
WTB11 |
344.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
|||||||||
17120335
|
Memmert™
WTB15D00123 |
WTB15 |
344.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
|||||||||
17120365
|
Memmert™
WTB24D00123 |
WTB24 |
344.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
|||||||||
17130425
|
Memmert™
WTB6D00123 |
WTB6 |
344.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
|||||||||
17150385
|
Memmert™
WTB35D00123 |
WTB35 |
344.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
|||||||||
17180405
|
Memmert™
WTB50D00123 |
WTB50 |
344.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
- Accessory
- Factory calibration certificate
- 160°C
Specifications
Works Calibration Certificate |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Memmert™ Works Calibration Certificate - Factory 160°C