missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XY 018
ROR/NR1F Antagonists
180.00 EUR - 665.00 EUR
Specifications
CAS | 1873358-87-2 |
---|---|
Synonym | N-[2′-Fluoro-4′-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl][1,1′biphenyl]-4-yl]-2-nitrobenzeneacetamide |
Molecular Formula | C23H15F7N2O4 |
Formula Weight | 516.36 |
Purity | 98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792458
|
Tocris Bioscience
6223/50 |
50 mg |
665.00 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773998
|
Tocris Bioscience
6223/10 |
10 mg |
180.00 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RORγ antagonist (EC50 = 190 nM). Suppresses androgen receptor expression in prostate cancer cell lines. Also inhibits growth of tumors in androgen receptor-positive xenograft models.Specifications
1873358-87-2 | |
C23H15F7N2O4 | |
98% |
N-[2′-Fluoro-4′-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl][1,1′biphenyl]-4-yl]-2-nitrobenzeneacetamide | |
516.36 |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
XY 018