Learn More
Eurofins DiscoverX YopH Protein, Active
Active, N-terminal GST-tagged recombinant Yersinia full length YopH, for use in Phosphatase Assays.
Brand: Eurofins DiscoverX 14-590M

Description
Essential virulence determinant. This protein is a protein tyrosine phosphatase. The essential function of YopH in Yersinia pathogenesis is host-protein dephosphorylation. It contributes to the ability of the bacteria to resist phagocytosis by peritoneal macrophages.
GenBank Y00551. The recombinant protein contains the amino acid substitution R211A with reference to Y00551. This mutation is reported inGenBank M30457.
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 10% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
250 μg | |
Recombinant | |
Recombinant enzyme expressed in E. coli cells | |
≥74% |
77.4 kDa | |
Yersinia species | |
N-terminal GST | |
Full-length | |
YopH, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.