Learn More
Eurofins DiscoverX YopH Protein, Active
Active, N-terminal GST-tagged recombinant Yersinia full length YopH, for use in Phosphatase Assays.

525.15 EUR - 4008.15 EUR
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 10% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 77.4 kDa |
Species | Yersinia species |
Recombinant | Recombinant |
Protein Tag | N-terminal GST |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17300394
![]() |
Eurofins DiscoverX
14-590 |
10 μg |
525.15 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17310394
![]() |
Eurofins DiscoverX
14-590M |
250 μg |
4008.15 EUR
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Essential virulence determinant. This protein is a protein tyrosine phosphatase. The essential function of YopH in Yersinia pathogenesis is host-protein dephosphorylation. It contributes to the ability of the bacteria to resist phagocytosis by peritoneal macrophages.
GenBank Y00551. The recombinant protein contains the amino acid substitution R211A with reference to Y00551. This mutation is reported inGenBank M30457.
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 10% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Yersinia species | |
N-terminal GST | |
Full-length | |
YopH, active |
77.4 kDa | |
Recombinant | |
Recombinant enzyme expressed in E. coli cells | |
≥74% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.