Learn More
BINDER™ Access Port
Access port with silicone plug for use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600.
Brand:  BINDER™ 8012-2488
 View more versions of this product
Product Code. 30238210
  • 943.00 EUR / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Description
  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600
Specifications
Specifications
KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600
Access Port
100 mm
Silicone
1 on Top
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Access Port > Access Port, 100 mm, Top