Learn More
Thermo Scientific™ DualSpin Fixed Angle Bucket for Medifuge Benchtop Centrifuge, set of 8
Use these buckets with the Medifuge Bench Top Centrifuge.
Brand: Thermo Scientific™ 75008815
154.67 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Use these buckets with the Medifuge Bench Top Centrifuge.
Specifications
Medifuge bench top centrifuge | |
Set of 8 | |
Fixed angle bucket |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%