Learn More
Invitrogen™ Human PHF15 Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP93570
Description
Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55275 (PA5-55275. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Scaffold subunit of some HBO1 complexes, which have a histone H4 acetyltransferase activity (PubMed:16387653). Acts as a E3 ubiquitin-protein ligase mediating the ubiquitination and subsequent proteasomal degradation of target protein histone demethylase KDM1A (PubMed:25018020). Also acts as a ubiquitin ligase E3 toward itself. Positive regulator of neurogenesis. [UniProt]
Specifications
Q9NQC1 | |
Blocking Assay, Control | |
23338 | |
100 μL | |
1200017K05Rik; AI480685; E3 ubiquitin-protein ligase Jade-2; jade family PHD finger 2; jade family PHD finger protein 2; JADE2; JADE-2; Kiaa0239; mKIAA0239; PHD finger protein 15; Phf15; protein Jade-2; protein Jade-2; E3 ubiquitin-protein ligase Jade-2; si:ch73-202g8.3 | |
JADE2 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human PHF15 Control Fragment | |
RUO | |
PHF15 | |
Unconjugated | |
Recombinant | |
ASVAADSDVQVLGPAASPKPLGRLRPPRESKVTRRLPGARPDAGMGPPSAVAERPKVSLHFDTETDGYFSDGEMSDSDVEAEDGGVQRGPREAGAEEVVRMGVLA | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.