Learn More
Macherey-Nagel™ NucleoSpin™ gDNA Clean-up, Mini kit for DNA clean up and concentration
Concentrate DNA samples more easily and quickly, compared to microdialysis filtrations units. Macherey-Nagel™ NucleoSpin™ gDNA Clean-up Kit cleans and concentrates large-sized genomic DNA from pre-purified DNA from organic extraction and from enzymatic extractions.
Brand: Macherey-Nagel™ 740230.250
Description
The NucleoSpin™ gDNA Clean-up kit is designed for the rapid purification of previously isolated small and especially high molecular weight genomic DNA. It is used to clean-up and concentrate genomic DNA after crude extraction methods, for example using Trizol, or after enzymatic, or chemical reactions. The time-saving procedures enables a preparation time of just 15 minutes for 10 preps.
- No need for organic denaturants or chloroform extractions
- Any impurities like phenol, enzymes, salts, dyes, labels, nucleotides, small oligonucleotides, and even up to 5% detergents (e.g., SDS, Triton, Tween, Lauroylsarcosin) are removed completely
- Binding Buffer DB and Wash Buffer DW are developed to allow a very gentle binding and washing
- High recovery rates up to 90% for high molecular weight DNA (up to 50 kbp) as well as for small DNA fragments down to 100 bp
- The eluted DNA is ready to use for downstream applications such as PCR, endonuclease restriction, Southern Blotting and labeling
- NucleoSpin™ gDNA Clean-up Columns, Collection Tubes (2 mL), Binding Buffer DB, Wash Buffer DW, Elution Buffer DE
Specifications
Mini Spin Column | |
DNA (Genomic) | |
Elution volume: 50–100μL | |
NucleoSpin™ gDNA Clean-up Columns (250), 2mL Collection Tubes (250), Binding Buffer DB (125mL), Wash Buffer DW Concentrate (3 x 50mL), Elution Buffer DE (30mL) | |
Nucleo Spin gDNA Clean Up | |
<150 μL solution containing <25 μg DNA |
Silica Membrane Technology | |
Recovery rate: 80–90% | |
Nucleo Spin gDNA Clean Up | |
250 | |
A260/A280: 1.8–1.9 | |
<15 min/10 preps |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.