Learn More
RAB38 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542210
Description
Peptide sequence: MQAPHKEHLY KLLVIGDLGV GKTSIIKRYV HQNFSSHYRA TIGVDFALKV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 90%
RAB38 Polyclonal antibody specifically detects RAB38 in Human samples. It is validated for Western Blot.
Specifications
RAB38 | |
Polyclonal | |
Unconjugated | |
Rab38 | |
2310011F14Rik; AU043391; chocolate; cht; melanoma antigen NY-MEL-1; NY-MEL-1; R; RAB38; Rab38, member of RAS oncogene family; RAB38, member RAS oncogene family; rab38a; RAB38a, member RAS oncogene family; Rab-related GTP-binding protein; ras-related protein Rab-38; red eyed dilution; rrGTPbp; Ruby; ruby or red eyed dilution; small GTP binding protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23682 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P57729 | |
Rab38 | |
Synthetic peptide directed towards the N-terminal of human RAB38 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.