Learn More
Fisherbrand™ S 50 R Ultrasonic Bath
Fisherbrand™ S 50 R Ultrasonic Water Bath features microprocessor-controlled ultrasonic modes and special predefined programs to assist every standard laboratory and cleaning application.
Brand:  Fisherbrand™ 1045437
Product Code. 12689345
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • Plastic carrying handles
  • Detachable mains cable
  • User-friendly clearly arranged operating panel
  • Connection rack for laboratory holder, noise-reduction operation
  • Ultrasonic tank made of cavitation-resistant stainless steel
  • Special efficient and noise-reduced Degas mode (gas liftout effect)
  • Turning knob for setting continued and short-period operation from 1 to 30 min
  • LED display for both pre-set and actual temperature, and for remaining operating time
Specifications
Specifications
230 VAC
335 x 304 mm (H x Dia.)
335 mm
Elmasonic S 50 R
5.3 L
230VAC, 150w
335 mm
150 w
Videos
  • Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
    Certificates
    View accessory products
    Product Title
    Fisherbrand™ S 50 R Ultrasonic Bath > Capacity: 5.3L