Learn More
Memmert™ Stainless Steel Bottom Drip Tray
15 mm stainless steel bottom drip tray
213.00 EUR - 469.00 EUR
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | |||||
17109764
|
Memmert™
HPP260ecoB29722 |
HPP260eco |
261.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
17109864
|
Memmert™
HPP750ecoB34055 |
HPP750eco | Only in connection with option K1 |
406.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
17119854
|
Memmert™
HPP750ecoB04362 |
HPP750eco | Not in connection with option K1 |
406.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
17139664
|
Memmert™
HPP110ecoB04359 |
HPP110eco | Not in connection with option K1 |
213.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
17160045
|
Memmert™
IPP1060ecoB29769 |
IPP1060eco | Not in connection with option K1 |
469.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
17160165
|
Memmert™
IPP260ecoB29722 |
IPP260eco |
261.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
17170245
|
Memmert™
IPP750ecoB04362 |
IPP750eco | Not in connection with option K1 |
406.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
17179624
|
Memmert™
HPP1060ecoB29769 |
HPP1060eco | Not in connection with option K1 |
469.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
17180205
|
Memmert™
IPP410ecoB29722 |
IPP410eco |
261.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
17189804
|
Memmert™
HPP410ecoB29722 |
HPP410eco |
261.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
17190075
|
Memmert™
IPP110ecoB04359 |
IPP110eco | Not in connection with option K1 |
213.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
Description
- May affect the temperature distribution
- 15 mm
Specifications
15 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Memmert™ Stainless Steel Bottom Drip Tray