missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Access Port

Access port with silicone plug for use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600.

Brand:  BINDER™ 8012-2485

 View more versions of this product

Product Code. 30238259

  • 589.00 EUR / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600
Specifications

Specifications

KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600
Access Port
50 mm
Silicone
1 on Right Side
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BINDER™ Access Port > Access Port, 50 mm, Right

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.