missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11444931
|
MP Biomedicals
0215574280 |
100 mg |
137.80 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454931
|
MP Biomedicals
0215574290 |
500 mg |
455.80 EUR
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464931
|
MP Biomedicals
0215574291 |
1 g |
733.52 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
84195-52-8 |
Specifications
84195-52-8 | |
≈400,0002 | |
Mucin |
Tan | |
Flaky Powder |
Safety and Handling
missing translation for 'einecsNumber' : 282-357-4
Recommended Storage : Store at 4°C, dessicate.